missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Amber Acid Storage Bottle Cap
Click to view available options
Diametro (sistema metrico):
48 mm
55 mm
66 mm
Descrizione
Made of chemically resistant amber DURAN™ glass. With standard taper pennyhead stopper and additional ground glass cap. This provides an improved seal and protection against acid vapours. Ambered glass provide protection from Ultra-violet light.
- Protection from UV light due to amber colouration
- Ground DURAN™ glass stopper and additional DURAN™ glass cap provides protection against acid vapours
- Pennyhead stopper with standard taper joint
- Conical shoulders
- Replacable glass cap
Specifica
Diametro (sistema metrico) | 66 mm |
Da utilizzare con (apparecchiatura) | For 500 mL bottle |
Descrizione | Cap for 500 mL Bottle |
Da utilizzare con (applicazione) | Storage and dispensing of acids; Storage of photosensitive reagents |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'