Learn More
DWK Life Sciences DURAN™ Bell Jar, with glass knob top
Flat flange bell with knob and ground flange for mounting on a vacuum experimental plate (vacuum chamber).
Marca: DWK Life Sciences 244606907
541.50 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 6.00000kg
Descrizione
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
- The button makes it easy to raise the bell jar
- Fired-on, highly durable white ceramic
Specifica
315 mm | |
Vacuum applications; Tests in vacuum chambers | |
Yes |
Borosilicate Glass (3.3) | |
1/Pk. |