Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks are manufactured from durable borosilicate glass.
Marca: Lenz Laborglasinstrumente™ B0418721
Dettagli aggiuntivi : Peso : 0.47700kg
Descrizione
- With inscription label
- Available in a range of capacities
- Corrosion-free and resistant to thermal shock
Specifica
2000 mL | |
Vetro borosilicato Duran™, Duran Glass | |
Head Touches NSH 45/40 | |
Rotondo |