Learn More
DWK Life Sciences DURAN™ Crystallizing Dish, without spout
Codice prodotto. 11746582 Sfoglia Tutto DWK Life Sciences Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
20 mL
40 mL
60 mL
100 mL
150 mL
300 mL
500 mL
900 mL
2000 mL
3500 mL
Dimensione della confezione:
pezzi 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) unitSize
11746582 20 mL pezzi 10
11756582 40 mL pezzi 10
11766582 60 mL pezzi 10
11776582 100 mL pezzi 10
11786582 150 mL pezzi 10
11746592 300 mL pezzi 10
11756592 500 mL pezzi 10
11766592 900 mL pezzi 10
11776592 2000 mL pezzi 10
15221689 3500 mL pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Codice prodotto. 11746582 Fornitore DWK Life Sciences N. del fornitore 213132403
Trusted Glassware Deals
Usa il codice promozionale 28509
Visualizza tutte le promozioni
Spedizione stimata: 04-08-2026
Aggiungi al carrello
Aggiungi al carrello
The flat shape as well as the flat bottom of the crystallizing dish allow an even evaporation of solvents and thus the crystallization of solids from solutions and suspensions.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Flat and wide shape allow solvents to evaporate quickly and evenly
  • Large ground base
  • Spout for clean pouring
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifica
Autoclavabile Autoclavable
Capacità (sistema metrico) 20 mL
Materiale Borosilicate Glass (3.3)
Da utilizzare con (applicazione) Crystallization; Preparation of concentrated solutions; Available with spout DIN 12 338 or without spout DIN 12 337
N. per confezione 10/Pk.
Diametro (sistema metrico) 40 mm