Descrizione
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
- Spout for clean pouring
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Capacità (sistema metrico) | 15 mL |
| Materiale | Borosilicate Glass (3.3) |
| Tipo | Without Labeling Field |
| Da utilizzare con (applicazione) | Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field |
| N. per confezione | 10/Pk. |
| Diametro (sistema metrico) | 50 mm |