Learn More
DWK Life Sciences DURAN™ Evaporating Flask, with standard ground joint, pear shape
Product Code. 15241859 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15241859 100 mL Pack of 10
15251859 250 mL Pack of 10
15261859 500 mL Pack of 10
15271859 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15241859 Supplier DWK Life Sciences Supplier No. 241202707
  • ONLINE EXCLUSIVE PRICE
    359.00€
    339.40€ / pezzi 10
    Risparmia 19.60€
    5% Sconto
View all promotions
Spedizione stimata: 11-12-2026
Evaporating flasks are particularly suitable for evaporating liquids or solvents.
Evaporating flasks are a variant of round flasks and have a pear-shaped shape. The standard ground joint allows a precise connection to other ground joint components, e.g. a rotary evaporator. Due to their shape, evaporating flasks allow a large uniform liquid surface, making them particularly suitable for the evaporation of solvents.
  • Tight closure due to standard ground joint
  • Fired-on, highly durable white print
  • Pear shape
Specifications
Description Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) Evaporation of liquids; Heating; Suitable for heating baths and mantles; Mixing several components; Rotary evaporator flask, suitable for all standard rotary evaporators; Concentration of solutions
Quantity 10/Pk.
Volume (Metric) 100 mL
Diameter (Metric) 60 mm