Learn More
DWK Life Sciences DURAN™ Filter Tube, Allihn type
Micro filter tubes can be used to filter very small quantities of solution. The Allihn design has a large volume (30 ml) compared to other microfilters.
382.00€ - 415.00€
Specifica
Materiale | Borosilicate Glass (3.3) |
---|---|
Da utilizzare con (applicazione) | Filter design according to Allihn; Filtration of very small quantities; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering |
N. per confezione | 10/Pk. |
Codice del prodotto | Marca | Descrizione | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Descrizione | Prezzo | Quantità e disponibilità | |||||
12526548
|
DWK Life Sciences
258532206 |
Porosity 2 |
382.00€
pezzi 10 |
Spedizione stimata: 02-07-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
15262049
|
DWK Life Sciences
258532309 |
Porosity 3 |
382.00€
pezzi 10 |
Spedizione stimata: 30-07-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
15272049
|
DWK Life Sciences
258532403 |
Porosity 4 |
415.00€
pezzi 10 |
Spedizione stimata: 30-07-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
15252049
|
DWK Life Sciences
258532103 |
Porosity 1 |
382.00€
pezzi 10 |
Spedizione stimata: 30-07-2024
Eseguire il login per visualizzare lo stock disponibile |
|||||
Descrizione
One of the most important separation methods in the laboratory environment is filtration, which is used to separate or clean different media. For this working method, filters are required which are selected according to the application with regard to size and porosity. Filter funnels are filters that are used for solid-liquid separation by means of vacuum. Filter frits have a glass sinter bottom which serves as a filter medium and does not require any further filter paper.
- Allihn type
- Suitable for filtration of very small quantities due to filter size
- Fired-on, highly durable white print
Specifica
Borosilicate Glass (3.3) | |
Filter design according to Allihn; Filtration of very small quantities; Filtration at reduced pressure; Qualitative inorganic analysis and preparative chemistry; Filtering | |
10/Pk. |