Learn More
Lenz Laborglasinstrumente™ Beute a tre colli in vetro DURAN™
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks are manufactured from durable DURAN™ glass.
Marca: Lenz Laborglasinstrumente™ B0424901
Dettagli aggiuntivi : Peso : 0.27000kg
Descrizione
- Acc. to DIN 12392
- With conical ground joints
- Parallel side necks
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifica
500 mL | |
Vetro Duran, Duran Glass |