Learn More
Lenz Laborglasinstrumente™ Beute a tre colli in vetro DURAN™
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Round Bottom Three-Neck Flasks are manufactured from durable DURAN™ glass.
Marca:  Lenz Laborglasinstrumente™ B0424901
Dettagli aggiuntivi : Peso : 0.27000kg
Codice prodotto. 10713631
  • 56.00€ / Pezzo
Spedizione stimata: 14-05-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Acc. to DIN 12392
  • With conical ground joints
  • Parallel side necks
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifica
Specifica
500 mL
Vetro Duran, Duran Glass