Learn More
DWK Life Sciences DURAN™ Membrane Venting GL Screw Cap, Blue PP, Welded-in ePTFE Membrane for Pressure Equalisation During Autoclaving
Sfoglia Tutto DWK Life Sciences ProdottiDescription
- Pressure equalisation during steam sterilisation, the 0.2μm ePTFE membrane permits automatic sterile gas exchange in two directions
- Microporous ePTFE membrane blocks ingress of liquids or solids, keeping bottle contents sterile throughout the autoclave cycle
- Increased gas exchange rate due to thin 0.2μm ePTFE membrane combined with larger ventilation openings
- PTFE membrane and polypropylene mesh support are ultrasonically welded in place, forming a durable seal
- Reusable and cost effective — replacement recommended after 3 to 5 autoclave cycles or if membrane becomes damaged or contaminated
- ePTFE membrane conforms to USP 121°C Class VI; supported by polypropylene mesh
- Compatible with GL 25, GL 32, and GL 45 DIN thread laboratory bottles; ideal for storage or transport of gas-generating media
Specifications
Specifications
| Colore | Blue |
| Da utilizzare con (applicazione) | Pressure equalisation during steam sterilisation/autoclaving of media or buffers; Sterile gas exchange for aerobic microbial, shaker and suspension cell cultures; Autoclaving of media and buffers; Short-term storage of sterilised media; Storage of gas generating materials |
| Materiale | Polypropylene, ePTFE |
| Quantità | 5/Pk. |
| Sterilità | Non-sterile |
| Dimensioni filettatura (sistema metrico) | GL 32 |
| Diametro (sistema metrico) | 41 mm |
| Autoclavabile | Autoclavable |
| Monouso | Reusable |
| Product Type | Bottle Cap |