Learn More
DWK Life Sciences DURAN™ Mixing Cylinder, with Hexagonal Base, Class B
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Capacità (sistema metrico):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
pezzi 2
Descrizione
DURAN™ Mixing cylinders are for diluting solutions and mixing several components in a given quantity ratio. The mixing cylinders of Class B have a line graduation and a white scale. Volume content tolerances for measuring and mixing cylinders conform to DIN and ISO accuracy limits.
- Calibration is based on the poured in volume (In) at a + 20°C reference temperature
- Large hexagonal base equipped with three knobs which increase its stability and prevent the cylinder from rolling
- A uniform wall thickness over the entire measuring range prevents wedge errors
- Supplied with octagonal polyethylene stopper
- Tight closure due to standard ground joint
- Fired-on, highly durable white print
- Easy-to-read scale with graduation marks to estimate the content
- The volume content tolerances conform to accuracy Class B
Specifica
Specifica
| Tolleranza | ± 0.5 mL |
| Materiale | Borosilicate Glass (3.3), Polyethylene |
| Capacità (sistema metrico) | 25 mL |
| Diameter (Metric) Outside | 21 mm |
| Da utilizzare con (applicazione) | Measuring liquid amounts; Diluting solutions; Mixing several components; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers |
| Include | 2/Pk. |
| Autoclavabile | Autoclavable |
| Temperatura max | 250°C |