Learn More
DWK Life Sciences DURAN™ Multi-purpose Cylinder, with round base, without graduation
Sfoglia Tutto DWK Life Sciences ProdottiDescrizione
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility.
- Rough ground rim
- With round base
Specifica
Specifica
| Materiale | Borosilicate Glass (3.3) |
| Diameter (Metric) Outside | 40 mm |
| Da utilizzare con (applicazione) | Diluting solutions; Mixing several components |
| Capacità (sistema metrico) | 180 mL |
| Autoclavabile | Autoclavable |
| Quantità | 10/Pk. |