Learn More
DWK Life Sciences DURAN™ NMR Tubes, for four application frequencies, with closures
Sfoglia Tutto DWK Life Sciences ProdottiDescrizione
Nuclear magnetic resonance spectroscopy (NMR-spectroscopy) is a spectroscopic method to elucidate the structure of molecules and compounds. Due to electronic interactions of single atoms with the neighbouring atoms, structural conclusions can be drawn. In addition to structure elucidation, the concentration and dynamics of molecules can be determined with this method. This analytical method requires very precise tubes with a constant diameter, a uniform wall thickness and excellent straightness across the entire tube.
- Close tolerances and accuracy, especially to their straightness, wall thickness and wall thickness distribution
- Supplied with cap
Specifica
Specifica
| Materiale | Borosilicate Glass (3.3), EVA |
| Product Type | NMR Tube |
| Diametro (sistema metrico) esterno | 4.97±0.013 mm |
| Forza del campo | 500 MHz |
| Da utilizzare con (applicazione) | Nuclear magnetic resonance spectroscopy (NMR spectroscopy); Storage and transport of measurement solutions; Designed for use in low to most mid field NMR spectrometers |
| Quantità | 5/Pk. |
| Modello | Scientific with Retrace Code |