Learn More
DWK Life Sciences DURAN™ Oil Centrifuge Tubes
Used for the determination of water and sediments in crude oils.
dietro preventivo
Specifica
Capacità (sistema metrico) 100 mL
Monouso Reusable
Materiale Borosilicate Glass (3.3)
Da utilizzare con Centrifugation, Separation of Different Particles, Separation of Materials, Petrochemistry
Sterilità Non-sterile
Visualizza altre specifiche
Descrizione
Descrizione
  • Markings and graduations are lasered and therefore have excellent resistance to aggressive solvents.
  • Unsurpassed legibility and line uniformity across the entire scale range
  • Complies with USP Type I and ASTM E438, Type I, Class A as well as the specifications of the American Petroleum Institute (API) and the American Society for Testing and Materials (ASTM).
  • Calibrated to contain
  • The closest volumetric measurement tolerance exists compared to competing products.
Specifica
Specifica
100 mL
Borosilicate Glass (3.3)
Non-sterile
Yes
Reusable
Centrifugation, Separation of Different Particles, Separation of Materials, Petrochemistry