Learn More
DWK Life Sciences DURAN™ PURE GL 45 Premium Pouring Ring
BIOPHARMA
Product Code. 16211832 Shop All DWK Life Sciences Products
Change view
Click to view available options
Material:
PFA
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
16211832 PFA Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16211832 Supplier DWK Life Sciences Supplier No. 891088678
  • ONLINE EXCLUSIVE PRICE
    19.68€
    23.21€ / pezzi 5
    Risparmia dietro preventivo
    -17.94% Sconto
In stock
Add to basket
Edge
Add to basket
DURAN™ PURE GL 45 Pouring ring for screw closures developed for the needs of the pharmaceutical and biotechnology industries.
  • Manufactured from PFA
  • For use with DURAN™ PURE GL 45 Screw Caps
Specifications
Color Colorless
For Use With (Application) Manufacturing of Pharmaceutical Drugs and Biotech Products; Storage and Transport of APIs, Bulk Drug Intermediates, Ingredients and Media
Quantity 5/Pk.
Material PFA
Type Pouring Ring