Learn More
DWK Life Sciences DURAN™ Rolled Flange Bottle, without closure
Click to view available options
Capacità (sistema metrico):
100 mL
250 mL
500 mL
1000 mL
Descrizione
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. While some bottles and flasks are used for the cultivation of micro-organisms, the roller rim bottles are mainly used for sampling (e.g. in the dairy industry).
- Supplied without screw cap
Specifica
Diametro (sistema metrico) | 57 mm |
Capacità (sistema metrico) | 250 mL |
Da utilizzare con (applicazione) | Sample collection; Storage and transport of Liquid cultures; Media preparation and cultivation; Delivery without closure |
Materiale | Borosilicate Glass (3.3) |
Quantità | 10/Pk. |
Autoclavabile | Autoclavable |