Learn More
DWK Life Sciences DURAN™ Watch Glass Dish, fused rim
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Diametro (sistema metrico):
40 mm
50 mm
60 mm
80 mm
100 mm
125 mm
150 mm
200 mm
250 mm
Dimensione della confezione:
Pezzo
pezzi 10
Descrizione
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Watch glasses, for example, are concave curved. The small trough created in this way allows liquids or solids to be filled in.
- Reduced risk of injury due to fused rim
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Materiale | Borosilicate Glass (3.3) |
| Da utilizzare con (applicazione) | Weighing; Covering; Pretreatments; Drying of materials; Available in Boro 3.3 or soda-lime glass |
| N. per confezione | 10/Pk. |
| Diametro (sistema metrico) | 100 mm |