missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves

Provides a stronger, economical alternative to household rubber gloves. Ansell Edmont™ Duratouch™ 34-755 Series Single Use Vinyl Gloves are ambidextrous single-use vinyl gloves.

Marca:  Ansell Edmont™ 34755075

Dettagli aggiuntivi : Peso : 0.54708kg

 vedi altre versioni di questo prodotto

Codice prodotto. 15339177

  • 11.35€ / pezzi 100


Spedizione stimata
12-04-2019
Aggiungi al carrello

Descrizione

Descrizione

  • Single use gloves
  • Reduces costs for hand/ product protection wherever changeover rate is high
  • No seams to leak or split
  • Liquid proof, with no point of weakness
  • Pre-powdered
  • More comfortable to wear and easier to put on and remove without tearing
  • Gloves contain no talcum powder which might contaminate products
  • Ambidextrous, fits either hand
  • More economical and convenient: no need to pair left and right

Applications:

  • Painting, coating and cleaning
  • Laboratory analysis
  • Food Processing
  • Agriculture
  • Maintenance
Specifica

Specifica

EN 420:2003 + A1:2009 and EN374:2003
Rolled beaded
100 gloves in box, 10 boxes in carton
7.5
Vinyl gloves
Trasparente
Smooth Finish
Vinyl
34-755
0.12mm
Single use gloves
240mm
Videos