Learn More
ABEBA™ Dynamic Flow Burgundy Occupational Slip On O1 ESD
Burgundy slip-on occupational shoes with replaceable active MediFit insole, slip resistant TPU outsole
Marca: ABEBA™ 37374/39
Dettagli aggiuntivi : Peso : 0.63000kg
Descrizione
- Breathable, lightweight occupational slip-on shoes
- Comply to EN ISO 20347: 2012, O1 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Burgundy knitted fabric breathable
- Double elastic band fitting system
- Removable active Medifit insole
Specifica
Burgundy | |
39 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340 | |
Slip-on | |
O1 SRC ESD |