Learn More
ABEBA™ Dynamic Flow Burgundy Occupational Slip On O1 ESD
Burgundy slip-on occupational shoes with replaceable active MediFit insole, slip resistant TPU outsole
Marca:  ABEBA™ 37374/41
Dettagli aggiuntivi : Peso : 0.73000kg
Codice prodotto. 17101395
  • 67.38€ / 1 paio
Spedizione stimata: 27-05-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Breathable, lightweight occupational slip-on shoes
  • Comply to EN ISO 20347: 2012, O1 SRC
  • ESD dissipative according EN 61340 class climatic 1
  • Slip resistant TPU outsole
  • Good cushioning
  • Energy absorption in heel area
  • Burgundy knitted fabric breathable
  • Double elastic band fitting system
  • Removable active Medifit insole
Specifica
Specifica
Burgundy
41
Unisex
Water Repellent
Knitted Fabric
Comply to EN ISO 20347: 2012 O1 SRC, ESD class climatic 1 according EN 61340
Slip-on
O1 SRC ESD