Learn More
ABEBA™ Dynamic White Occupational Clogs Ob ESD
White dynamic occupational clogs with non-foldable heel strap, replaceable active comfort insole, slip resistant TPU outsole
Marca: ABEBA™ 37329/47
Dettagli aggiuntivi : Peso : 0.80800kg
Descrizione
- Breathable, lightweight, comfortable occupational clogs
- Comply to EN ISO 20347: 2012, OB SRC
- ESD dissipative according to DIN EN 61340
- Slip resistant TPU outsole
- Good cushioning
- Energy absorption in heel area
- Water repellent white knitted textile
- Non foldable heel strap, adjustable
- Removable active comfort insole
Specifica
White | |
47 | |
Unisex | |
Water Repellent |
Knitted Fabric | |
ESD dissipative to EN 61340 Climate class 1, Comply to EN ISO 20347: 2012 OB SRC | |
Clogs | |
OB SRC ESD |