missing translation for 'onlineSavingsMsg'
Learn More

DYNC2LI1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA564075

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15751842

  • 514.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: PENDIGKLHA HSPMELWKKV YEKLFPPKSI NTLKDIKDPA RDPQYAENEV DEMRIQKDLE LEQYKRSSSK SWKQIEL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 86%.

DYNC2LI1 Polyclonal antibody specifically detects DYNC2LI1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifica

Specifica

DYNC2LI1
Polyclonal
Unconjugated
Dync2li1
4933404O11Rik; CGI-60; Cytoplasmic dynein 2 light intermediate chain 1; D2LIC; Dync2li1; Dynein 2 light intermediate chain; dynein cytoplasmic 2 light intermediate chain 1; dynein, cytoplasmic 2, light intermediate chain 1; LIC3; mD2LIC; RGD1310286
Rabbit
Antigen affinity chromatography
RUO
51626
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.9 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8TCX1
Dync2li1
Recombinant protein corresponding to Human DYNC2LI1
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only