Learn More
3M™ E-A-R™ UltraTech Earplugs
Codice prodotto. 15865985
Cambia vista
Click to view available options
:
Color: Black, White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
15865985 Color: Black, White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15865985 Fornitore 3M™ N. del fornitore 7000103799
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
3M™ E-A-R™ UltraTech Earplugs greatly improve the ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively.
  • Improve wearer's ability to perceive speech, warning signals and machinery noise whilst reducing harmful noise levels effectively
  • Earplug are ideal for musicians who require a flat attenuation protection
  • Conical shape plugs help provide effective seal for reassured protection
  • Effective protection against moderate levels of harmful noise SNR 21 dB
TRUSTED_SUSTAINABILITY
Specifica
Colore Black, White
Pairs per Package 1 Pair/Box
Noise Reduction Rating (NRR) 21 dB
Corded Uncorded
Dielectric No