Learn More
3M™ E.V.P.™ Tactical Protective Eyewear Prescription Lens Insert 40719-00000
Sfoglia Tutto 3M ProdottiDescrizione
- For polycarbonate prescription lenses up to + and - 6 diopter
- This lens is helpful for anyone who needs corrective vision to go along with eye protection
- Insert available for 3M™ Maxim™ Ballistic Safety Glasses
Specifica
Specifica
| Certificazione/Conformità | CE Approved, EN 166:2001 |
| Lens Coating | Scratch-resistant |
| Descrizione | Light Amber Lens Color |
| Lens Tint | Amber |
| Product Type | Lens Insert |
| Colore telaio | Clear, Gray |