Learn More
Hanna Instruments™ Edge™ conductivity and temperature probe, digital, four-ring, platinum, HI763100, Hanna Instruments
Product Code. 15138249
Change view
Click to view available options
Material:
PEI Body, Platinum Tip
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
15138249 PEI Body, Platinum Tip Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15138249 Supplier Hanna Instruments™ Supplier No. HI763100
  • ONLINE EXCLUSIVE PRICE
    1179.75€
    744.62€ / pezzi 1
    Risparmia 435.13€
    36% Sconto
Spedizione stimata: 30-11-2026
Specifications
Type Digital Conductivity Electrode
Includes Temperature Sensor
Material PEI Body, Platinum Tip
Temperature (English) Operating 23°F to 212°F
Temperature (Metric) Operating -5°C to +100°C
Form 4-ring Probe