missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF2 beta Polyclonal Antibody
Descrizione
Immunogen sequence: PDEDEILEKD EALEDEDNKK DDGISFSNQT GPAWAGSERD YTYEELLNRV FNIMREKNPD MVAGEKRKFV MKPPQVVRVG TKKTSFVNFT DICKLLHRQP KHLLAFLLAE LGTSGSIDGN NQLVIKGRFQ QKQIENVLRR YIKEYVTCHT CRSPDTILQK DTRLYFLQCE TCHSRCSVAS IKTGFQAVTG KRAQLRAKAN; Positive Samples: HeLa, SKOV3, HepG2, Mouse heart, Mouse spinal cord.
Eukaryotic translation initiation factor 2 (EIF-2) functions in the early steps of protein synthesis by forming a ternary complex with GTP and initiator tRNA and binding to a 40S ribosomal subunit. EIF-2 is composed of three subunits, alpha, beta, and gamma, with the protein encoded by this gene representing the beta subunit. The beta subunit catalyzes the exchange of GDP for GTP, which recycles the EIF-2 complex for another round of initiation. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifica
Specifica
| Antigene | eIF2 beta |
| Applicazioni | ELISA, Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.38 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EIF2S2 |
| N. accesso geni | P20042, Q99L45 |
| Alias gene | 2810026E11Rik; 38kDa; AA408636; AA571381; AA986487; AW822225; D2Ertd303e; EIF2; EIF2B; EIF2BA; EIF2beta; eIF-2-beta; EIF2S2; Eukaryotic translation initiation factor 2 subunit 2; eukaryotic translation initiation factor 2 subunit beta; eukaryotic translation initiation factor 2, subunit 2 (beta); eukaryotic translation initiation factor 2, subunit 2 (beta, 38kDa); eukaryotic translation initiation factor 2, subunit 2 beta; eukaryotic translation initiation factor 2, subunit 2 beta, 38kDa; PPP1R67; protein phosphatase 1, regulatory subunit 67 |
| Simboli geni | EIF2S2 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?