Learn More
EIF3M Antibody (1A4) [Allophycocyanin], Novus Biologicals Biologicals™
Codice prodotto. 30519921 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30519921 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30519921 Fornitore Novus Biologicals N. del fornitore NBP342965APC
Mouse Monoclonal Antibody
EIF3M Monoclonal antibody specifically detects EIF3M in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifica
Antigene EIF3M
Applicazioni ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classificazione Monoclonal
Clone 1A4
Coniugato APC
Formulazione PBS
Alias gene B5 receptor, dendritic cell protein, eIF3m, eukaryotic translation initiation factor 3 subunit M, eukaryotic translation initiation factor 3, subunit M, Fetal lung protein B5, FLJ29030, GA17, hfl-B5, HFLB5, PCI domain containing 1 (herpesvirus entry mediator), PCI domain-containing protein 1, PCID1B5
Specie ospite Mouse
Immunogeno eukaryotic translation initiation factor 3, subunit M - (Uniprot# Q7L2H7)
Metodo di purificazione Protein A or G purified
Quantità 0.1 mL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 10480
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG1