Learn More
Brady™ Electrical Lockout Kit
Product Code. 12718009 Shop All Brady Products
Change view
Click to view available options
Color:
Black/Blue/Red on White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12718009 Black/Blue/Red on White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12718009 Supplier Brady™ Supplier No. 805909
  • ONLINE EXCLUSIVE PRICE
    340.00€
    310.48€ / pezzi 1
    Risparmia 29.52€
    8% Sconto
In stock
Our Electrical Lockout Kit gives you all the basics to lock out breakers, electrical plugs, wall switches and fuses.
  • Our electrical lockout kit gives you all the basics to lock out breakers, electrical plugs, wall switches and fuses.
Specifications
Product Type Electrical Lockout Kit
Color Black/Blue/Red on White
For Use With (Application) Lockout Tagout
Quantity 1 Ea.