Learn More
Macherey-Nagel™ Electronic High Power Crimping Tool
Product Code. 12868595 Shop All Macherey-Nagel Products
Change view
Click to view available options
Size:
11 mm, 20 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12868595 11 mm, 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12868595 Supplier Macherey-Nagel™ Supplier No. 735700
In stock
  •  Crimping and decaping heads are purchased separately
Specifications
No. per Pack 1 Each
Operation Automated
Type Electronic High Power Crimping Tool
For Use With (Equipment) For all 11mm and 20mm Crimp Caps
Size 11 mm, 20 mm