Learn More
Bibby Scientific™ Electrothermal™ Replacement Spare Heater Element
Product Code. 11778422
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11778422 500 mL Each
11758422 100 mL Each
11768422 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11778422 Supplier Bibby Scientific™ Supplier No. 9996699
  • ONLINE EXCLUSIVE PRICE
    411.00€
    350.48€ / pezzi 1
    Risparmia 60.52€
    14% Sconto
Spedizione stimata: 17-12-2026
Specifications
Capacity (Metric) 500 mL
For Use With EMEA3/6/0500
Voltage 230 V
Wattage 200w
Product Type Replacement Spare Heater Element
Frequency 50 Hz