Learn More
Kartell Embedding Rings
Product Code. 18028031
Change view
Click to view available options
Color:
Blue
Green
Pink
Yellow
Unit Size:
Pack of 2000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
18028031 Blue Pack of 2000
18038041 Yellow Pack of 2000
18038031 Green Pack of 2000
18058041 Pink Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18028031 Supplier Kartell Supplier No. 0291904
  • ONLINE EXCLUSIVE PRICE
    178.00€
    142.76€ / pezzi 2000
    Risparmia 35.24€
    19% Sconto
 Restricted Item
Spedizione stimata: 11-12-2026
Embedding rings made from ABS plastic.
  • Embedding supports suitable for the processing and the histological inclusion of paraffin pieces
  • The inclusion rings can use the same microtome support (instrument used to make the histological sections of samples) of the inclusion cassettes
  • They can be set up on every kind of microtome thanks to their structure and to the possibility to obtain uniform paraffin blocks.
  • Embedding rings is an easy-filling and, after cutting, is always ready for following sections
Specifications
Product Type Embedding Ring
Material Plastic
Color Blue
Length (Metric) 41 mm
Width (Metric) 28 mm
Length (English) 1.61 in.
Width (English) 1.1 in.
Depth (Metric) 11 mm
Depth (English) 0.43 in.
Disposable Single-use