Descrizione
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifica
Specifica
| Da utilizzare con | Millipore Filtration Systems |
| Include | Adapter, male M Luer-Lok, nylon |
| Materiale | Nylon |
| Sterilità | Non-sterile |
| Tipo | Laboratory Filter Holders or Cyclones |
| Materiale involucro | Nylon |