missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories, and Replacement Parts for In-Line Filter Holder

Accessory and replacements for 47mm In-Line Filter Holder

Marca:  Merck XX4304702

Dettagli aggiuntivi : Peso : 0.10000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 10789201

  • 209.00€ / pezzi 6
Spedizione stimata: 11-06-2024
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Accessory and replacements for 47mm In-Line Filter Holder

Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifica

Specifica

Not Autoclavable
O-ring
47 mm In-line filter holder
Non-sterile
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali