Learn More
Merck Millipore Microbiological Analysis Membrane Filters, Gridded
Optimized for MF method microbiological analysis of water or other liquids
Marca: Merck Millipore HAWG047A0
Dettagli aggiuntivi : Peso : 0.02000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Descrizione
- MF-Type (mixed cellulose esters) filter membranes
- 0.45μm (Type HA) designed to give complete retention and maximum recovery of total coliforms and fecal coliform bacteria
- 0.7μm (Type HC) ideal for fecal coliform analyses
- Use 0.22μm membranes where 0.45μm pore size is not sufficient
- For beverage analysis, choose membranes with 0.8μm pore size
Compliance
Certified to meet Standard Methods for the Examination of Water and Wastewater requirements.
Specifica
Certified to meet standard methods for the examination of Water and wastewater requirements | |
Absorbent pads | |
Non-sterile | |
4.7cm | |
AO Pak Membrane Filter |
White | |
0.45μm | |
Grid | |
MF method microbiological analysis of water or other liquids |