missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Montage™ Gel Extraction Kit
Elutes DNA from agarose gels in 10 minutes
Marca: Merck LSKGEL050
Dettagli aggiuntivi : Peso : 0.80000kg
Includes: 50 gel nebulizers, 50 Ultrafree-MC 0.45μm separators, 50 microcentrifuge vials, 500mL stock of 50X modified TAE buffer
Descrizione
- Typically recovers >70% of 100-1000 bp DNA from standard agarose gels
- Converts agarose to a spray from which DNA is separated via a 0.45μm Ultrafree™-MC filter
- Prepares gel-purified PCR products for sequencing or cloning without need for further purification
An Amicon™ Bioseparations product.
Specifica
50 Gel nebulizers, 50 Ultrafree™-MC 0.45μm separators, 50 microcentrifuge vials, 500mL stock of 50X modified TAE Buffer | |
50 Pack |