Descrizione
- Accepts deep well or standard receiver plates.
- Suitable for use with manual or automated protocols.
- ANSI/SBS compliant footprint allows for easy robotic deck integration.
- Easily configured for both flow-to-waste and analyte collection.
- DirectStack™ technology for crosstalk-free filtrate collection.
- Manifold components are resistant to solvents.
Specifica
Specifica
| Monouso | Single-use |
| Tipo di filtro | Manifold |
| Include | Manifold Base, Standard Collar, Gaskets And Gasket Inserts, Tubing, Valves, Pressure Gauge |
| Tipo | Vacuum Manifold |