Learn More
Merck Millipore Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Marca: Merck Millipore MHA00P2TT
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
- Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
- Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
- Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifica
Amber | |
2-10°C | |
United States | |
50 x 2mL | |
Standard Plate Broth |
Total count | |
Non selective medium used to detect total heterotrophic microorganisms in Water and other liquids, TTC is used as an indicator for easy enumeration | |
1 Year | |
Liquid | |
Ampule |