Learn More
Merck Millipore Nutrient Media
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
Marca:  Merck Millipore MHA000P2F
Dettagli aggiuntivi : Peso : 0.80000kg
Codice prodotto. 11923886
  • 104.00€ / pezzi 50
Spedizione stimata
30-04-2021
Aggiungi al carrello
Descrizione
Descrizione
Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs
  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures
Order Info
Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.
Specifica
Specifica
Deep Red
Media
Testing waste and effluent Waters for fecal Coliforms
1 Year
Liquid
Ampule
Fecal Coliforms
2-10°C
United States
50 x 2mL
mFC Fecal Broth