Learn More
Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for the iCAP™ Series ICP-OES Systems
Product Code. 12794868 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
EMT Duo Torch
For Use With (Equipment):
iCAP 7000 Series
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type For Use With (Equipment) Unit Size
12794868 EMT Duo Torch iCAP 7000 Series Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12794868 Supplier Thermo Scientific™ Supplier No. 842312051841
 Restricted Item
Spedizione stimata: 30-12-2026
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance plasma torch for iCAP™ Series ICP-OES systems.
Enable simple, routine maintenance through the unique design of the Thermo Scientific™ Enhanced Matrix Tolerance (EMT) Plasma Torch for Thermo Scientific™ iCAP™ Series ICP-OES systems. The torch clips in and can be removed in just seconds.
Enhanced performance through advanced accessories We offer a dedicated torch designs for radial and axial plasma views, including:
  • EMT Radial Torch for radial-only systems
  • EMT Duo Torch for radial and axial (duo) view systems
Specifications
For Use With (Equipment) iCAP 7000 Series
Type EMT Duo Torch