Learn More
Ark-Plas Products™ Enlarging Check Valve
Created from Kynar. Ark-Plas Products™ Enlarging Check Valve is excellent for prevention of backflow of fluid. The valve allows for an increase in pressure of the fluid.
Marca:  Ark-Plas Products™ HDX59-DF0
Dettagli aggiuntivi : Peso : 0.04590kg
Codice prodotto. 11841180
  • 143.21€ / pezzi 10
In stock, pronto per la spedizione: 9
Aggiungi al carrello
Descrizione
Descrizione
  • Created from Kynar polypropylene
  • Ideal for preventing backflow of fluid
Specifica
Specifica
Natural
Kynar
1/8 x 3/8 in.