Learn More
Memmert™ Entry Port Right Centre/Top
For introducing connections at the side and can be closed by flap
Marca: Memmert™ HPP1060ecoF3
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Standard position right centre/top
- Not for modelsize 110 with light module
Specifica
Clear | |
HPP1060eco | |
23 mm |