Learn More
Enzymaster Enzyme Screening Kit
Product Code. 17864621
Change view
Click to view available options
For Use With (Application):
Enzyme screening of aldolases
Enzyme screening of iminereductases
Enzyme screening of ketoreductases
Enzyme screening of transaminases
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17864621 Enzyme screening of ketoreductases Each
17874621 Enzyme screening of iminereductases Each
17884621 Enzyme screening of aldolases Each
17894621 Enzyme screening of transaminases Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17864621 Supplier Enzymaster Supplier No. 200100
  • ONLINE EXCLUSIVE PRICE
    4499.63€
    3950.00€ / pezzi 1
    Risparmia 549.63€
    12% Sconto
 Restricted Item
Spedizione stimata: 30-11-2026
The Enzyme Screening Kit contains a wide range of wild type and engineered enzymes with a broad substrate scope for your screening!
  • Ketoreductase Kit
    • Enzyme kit containing 20 ketoreductases (E.C. 1.1.1.) for a quick screening
    • Applicable for ketone reduction and alcohol oxidation
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • IRED Kit
    • Enzyme kit containing 16 iminereductases (E.C. 1.5.1.) for a quick screening
    • Applicable for imine reduction and reductive amination
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Alolase Kit
    • Enzyme kit containing 20 aldolases (E.C. 4.1.2.) for a quick screening
    • Contains threonine, phenylserine and serine aldolases
    • Applicable for reversible aldol reaction of glycine and an aldehyde to β-hydroxy-α-l-amino acid
    • Diverse range of substrates possible, high selectivity
    • 0.5 g per enzyme
    • Formulated as lyophilized cell-free extract (CFE)
  • Transaminase Kit
    • Enzyme kit containing 21 transaminases (E.C. 2.6.1.) for a quick screening
    • Applicable for reversible transamination of an amino group from an amino donor to a keto amino acceptor
    • Diverse range of substrates possible, both (S)- and (R)-selective enzymes included
    • 0.5 g enzyme powder per vial
    • Formulated as lyophilized cell-free extract (CFE)
Specifications
Product Type Enzyme Screening Kit
For Use With (Application) Enzyme screening of ketoreductases
Content And Storage 20 vials with 0.5 g enzyme (CFE)