Learn More
EPA Method 515.3 Independent Check Standard Mixture 407 10-100 μg/mL in Methyl tert Butyl Ether, Dr. Ehrenstorfer
Product Code. 30320715
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30320715 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30320715 Supplier Dr. Ehrenstorfer Supplier No. DREA50000407MB
 Restricted Item
Spedizione stimata: 08-01-2027
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4,5-T-methyl ester∣2,4-DB-methyl ester∣2,4-D-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣4-Nitroanisole∣Acifluorfen methyl ester∣Bentazone-methyl∣Chloramben methyl ester∣Chlorthal-dimethyl∣Dalapon-methyl ester∣Dicamba-methyl ester∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Fenoprop-methyl ester∣Pentachloroanisole∣Picloram-methyl ester
CAS 1928-37-6, 18625-12-2, 1928-38-7, 2905-67-1, 100-17-4, 50594-67-7
Analyte or Component Names 2,4,5-T-methyl ester∣2,4-DB-methyl ester∣2,4-D-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣4-Nitroanisole∣Acifluorfen methyl ester∣Bentazone-methyl∣Chloramben methyl ester∣Chlorthal-dimethyl∣Dalapon-methyl ester∣Dicamba-methyl ester∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Fenoprop-methyl ester∣Pentachloroanisole∣Picloram-methyl ester
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Pentachloroanisole 10 μg/mL∣2,4,5-T-methyl ester 25 μg/mL∣Fenoprop-methyl ester 25 μg/mL∣3,5-Dichlorobenzoic acid methyl ester 50 μg/mL∣Acifluorfen methyl ester 50 μg/mL∣Chloramben methyl ester 50 μg/mL∣Dicamba-methyl ester 50 μg/mL∣2,4-DB-methyl ester 100 μg/mL∣2,4-D-methyl ester 100 μg/mL∣4-Nitroanisole 100 μg/mL∣Bentazone-methyl 100 μg/mL∣Chlorthal-dimethyl 100 μg/mL∣Dalapon-methyl ester 100 μg/mL∣Dichlorprop-methyl ester 100 μg/mL∣Dinoseb methyl ether 100 μg/mL∣Picloram-methyl ester 100 μg/mL