missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 525 PAH Mixture 750 500 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Specifica
Specifica
| Materiale o nome chimico | Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene |
| CAS | 208-96-8, 120-12-7, 56-55-3, 207-08-9, 50-32-8, 205-99-2 |
| Nomi componenti o analita | Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene |
| Shipping Condition | -18°C |
| Certificazione/Conformità | ISO/IEC 17025 |
| Solvent or Matrix | Dichloromethane |
| Quantità | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acenaphthylene 500 μg/mL∣Anthracene 500 μg/mL∣Benz[a]anthracene 500 μg/mL∣Benzo(k)fluoranthene 500 μg/mL∣Benzo[a]pyrene 500 μg/mL∣Benzo[b]fluoranthene 500 μg/mL∣Benzo[ghi]perylene 500 μg/mL∣Chrysene 500 μg/mL∣Dibenzo(a,h)anthracene 500 μg/mL∣Fluorene 500 μg/mL∣Indeno[1,2,3-cd]pyrene 500 μg/mL∣Phenanthrene 500 μg/mL∣Pyrene 500 μg/mL |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?