Learn More
EPA Method 603/8030, VOA Vial Preserved to pH 4 - 5, 72/cs, Dr. Ehrenstorfer
Specifica
Specifica
| Materiale o nome chimico | Sodium Phosphate Monobasic Anhydrous |
| CAS | 7558-80-7 |
| Nomi componenti o analita | Sodium Phosphate Monobasic Anhydrous |
| Shipping Condition | Room Temperature |
| Formula molecolare | H2NaO4P |
| Certificazione/Conformità | ISO 17025 |
| Sinonimo | Dihydrogen Monosodium Phosphate, Dihydrogen Sodium Phosphate, Monobasic Sodium Phosphate, Monobasic Sodium Phosphate (NaH2PO4), Monosodium Dihydrogen Orthophosphate, Monosodium Hydrogen Phosphate, Monosodium Phosphate, Sodium Dihydrogen Monophosphate, Sodium Dihydrogen Orthophosphate, Sodium Dihydrogen Phosphate, Sodium Dihydrogen Phosphate (NaH2PO4), Sodium Hydrogen Phosphate (NaH2PO4), Sodium Monobasic Phosphate, Sodium Monobasic Phosphate (NaH2PO4), Sodium Orthophosphate Monobasic, Sodium Phosphate (Na(H2PO4)), Sodium Phosphate, Monobasic, Sodium primary phosphate |
| SMILES | [O-]P(O)(O)=O.[Na+] |
| Industry Type | Environmental Testing |
| Molecular Weight (g/mol) | 119.98 |