missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 Internal Standard Mixture 4000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Codice prodotto. 30313143
Cambia vista
Click to view available options
Quantità:
5 x 1 mL
Dimensione della confezione:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30313143 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30313143 Fornitore Dr. Ehrenstorfer N. del fornitore DREYS09000005DI

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico 1,4-Dichlorobenzene-D4∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
CAS 3855-82-1, 15067-26-2, 1719-03-5, 1146-65-2, 1520-96-3, 1517-22-2
Nomi componenti o analita 1,4-Dichlorobenzene-D4∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10
Shipping Condition Room Temperature
Certificazione/Conformità ISO 17034
Solvent or Matrix Dichloromethane
Industry Type Food Testing
Quantità 5 x 1 mL
Concentration or Composition (by Analyte or Components) 1,4-Dichlorobenzene-D4 4000 μg/mL∣Acenaphthene D10 4000 μg/mL∣Chrysene D12 4000 μg/mL∣Naphthalene D8 4000 μg/mL∣Perylene D12 4000 μg/mL∣Phenanthrene D10 4000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.