Learn More
EPB41L4B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560150
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: PLHININKAE EKKVSEKTLQ TPLLPSPVAD HVKCNILKAQ LENASRVNIQ GGKEESPFVN INKKSSLQDA SVRSPIPIRV ETAQPAVEKP EIKPPRVRKL TRQYSFDEDD LPP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.
Up-regulates the activity of the Rho guanine nucleotide exchange factor ARHGEF18. Involved in the regulation of the circumferential actomyosin belt in epithelial cells (PubMed:22006950). Promotes cellular adhesion, migration and motility in vitro and may play a role in wound healing (PubMed:23664528). May have a role in mediating cytoskeletal changes associated with steroid-induced cell differentiation (PubMed:14521927). [UniProt]Specifica
EPB41L4B | |
Polyclonal | |
Unconjugated | |
EPB41L4B | |
6430543G08Rik; AA589614; AU021054; band 4.1-like protein 4B; BB007528; CG1; D4Ertd346e; Ehm2; Epb4.1l4b; EPB41L4B; erythrocyte membrane protein band 4.1 like 4B; erythrocyte protein band 4.1 like 4b; erythrocyte protein band 4.1-like 4b; erythrocyte protein band 4.1-like 4b; expressed in high-metastatic cells; expressed in high-metastatic cells; FERM-containing protein CG1; LULU2; Protein EHM2; RGD1562988 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54566 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H329 | |
EPB41L4B | |
Recombinant protein corresponding to Human EPB41L4B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only