missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Epigallocatechin, MedChemExpress
Description
(-)-Epigallocatechin (Epigallocatechin) is the most abundant flavonoid in green tea, can bind to unfolded native polypeptides and prevent conversion to amyloid fibrils.
Specifications
Specifications
| Materiale o nome chimico | (-)-Epigallocatechin |
| CAS | 970-74-1 |
| Colore | White |
| Note sul grado di purezza | Research |
| Formula molecolare | C15H14O7 |
| Quantità | 5 mg |
| Sinonimo | Epigallocatechin L-Epigallocatechin |
| Informazioni di solubilità | DMSO : ≥ 150 mg/mL (489.76 mM) |
| SMILES | O[C@H]1[C@@H](C2=CC(O)=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C3C1 |
| Molecular Weight (g/mol) | 306.27 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?