Learn More
Eppendorf™ HeatSealer S100 Accessory, Plate Adapters
Codice prodotto. 15713155 Sfoglia Tutto Eppendorf Prodotti
Cambia vista
Click to view available options
Product Type:
High Profile Adapter
Low Profile Adapter
PCR Plate Adapter
Dimensione della confezione:
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
15713155 PCR Plate Adapter Pezzo
15723155 High Profile Adapter Pezzo
15703155 Low Profile Adapter Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 15713155 Fornitore Eppendorf™ N. del fornitore 5391070026
In stock
For use with HeatSealer S100
  • Hermetic sealing of multiwell plates
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
Warranty
Un anno
TRUSTED_SUSTAINABILITY
Specifica
Wattage 320 W
Da utilizzare con (apparecchiatura) HeatSealer S100
Intervallo temperatura sigillatura 170°C
Product Type PCR Plate Adapter