Learn More
EPTC, TRC
Codice prodotto. 30383112
Cambia vista
Click to view available options
Quantità:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30383112 50 mg
30380650 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30383112 Fornitore Toronto Research Chemicals N. del fornitore TRCE59225350MG
 Articolo soggetto a restrizioni
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 759-94-4
Formula molecolare C9 H19 N O S
Molecular Weight (g/mol) 189.32
Sinonimo Carbamothioic acid, N,N-dipropyl-, S-ethyl ester, Carbamic acid, dipropylthio-, S-ethyl ester (6CI,8CI), Carbamothioic acid, dipropyl-, S-ethyl ester (9CI), Alirox, EPTC, EPTC (herbicide), Eptam, Eptam 6E, FDA 1541, NSC 40486, Niptan, R 1608, S-Ethyl dipropylthiocarbamate, Stauffer R 1608, Torbin
IUPAC Name S-ethyl N,N-dipropylcarbamothioate
SMILES CCCN(CCC)C(=O)SCC
Specifica
Materiale o nome chimico EPTC
Quantità 50 mg
InChI Formula InChI=1S/C9H19NOS/c1-4-7-10(8-5-2)9(11)12-6-3/h4-8H2,1-3H3
Peso formulazione 189.1187