missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EquiPhi29™ DNA Polymerase

Codice prodotto. 16279981
Cambia vista
Click to view available options
Quantità:
1000 U
5000 U
Dimensione della confezione:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
16279981 5000 U 1 set
17360680 1000 U -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16279981 Fornitore Thermo Scientific™ N. del fornitore A39392
Follow the Workflow - Explore the Savings
Usa il codice promozionale 27967
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Thermo Scientific™ EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution.

Thermo Scientific™ EquiPhi29 DNA Polymerase is a proprietary phi29 DNA polymerase mutant developed through in vitro protein evolution. This enzyme is significantly improved over phi29 DNA polymerase in protein thermostability, reaction speed, product yield, and amplification bias, while retaining all the benefits of the wild-type enzyme.

EquiPhi29 DNA Polymerase possesses strong strand displacement activity and 3'→5' proofreading exonuclease activity that acts preferentially on single-stranded DNA or RNA.

Features of EquiPhi29 DNA Polymerase:

  • Highest processivity and strand displacement activity—more than 70 kb long DNA stretches can be synthesized
  • Low amplification bias
  • Extremely high yields of amplified DNA, even from small amounts of template
  • Highly accurate DNA synthesis in a short time

Applications:
EquiPhi29 DNA Polymerase may be used in a wide variety of applications including:

  • Unbiased whole genome amplification (WGA)
  • Rolling circle amplification (RCA)
  • Protein-primed DNA amplification
  • Cell-free cloning of lethal DNA
  • In situ genotyping with padlock probes
  • RNA-primed DNA amplification

Notes:

  1. The addition of pyrophosphatase to the reaction mixture with EquiPhi29 DNA Polymerase may further enhance DNA synthesis
  2. Because of the enzyme's 3'→5' proofreading exonuclease, 3'-modified primers are highly recommended.
  3. EquiPhi29 DNA Polymerase can amplify DNA in a wide range of temperatures (30–45°C), with an optimal reaction temperature of 42°C.
TRUSTED_SUSTAINABILITY

Specifica

Product Type Stand-alone enzyme
Da utilizzare con (applicazione) MDA-WGA, RCA
Polymerase EquiPhi29 DNA Polymerase
Quantità 5000 U
Content And Storage

• 5 x 100 μL EquiPhi29 DNA Polymerase (10 U/μL)
• 3 x 1 mL EquiPhi29 DNA Polymerase Reaction Buffer (10X)
• 0.25 mL DTT (100 mM)

Store at –15 to –25°C.

Optimal Reaction Temperature 42°C
Shipping Condition Dry Ice
Concentrazione 10 U/μL
Tempo di reazione 2 hours

For Research Use Only. Not for use in diagnostic procedures.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.